Learn More
Jeio Tech Inc Jeio Tech™ ON4-15V, 147L Advanced Natural Convection (Gravity) Oven (120V, 60Hz), PID Temp. Control, Gentle Uniform Heating, Temperature Ramping, Data Logging (RS-232/USB)
SDP
Catalog No. NC3965881
Encompass_Preferred
Jeio Tech ON4-15V Natural Convection (Gravity) Oven is engineered for gentle, reliable thermal processing with uniform temperature distribution, ideal for drying heat-sensitive materials, powders, and glassware without airflow disturbance. Utilizing natural gravity convection, the PID control system delivers excellent stability (±0.5°C fluctuation) and uniformity (±3.5°C variation) across a temperature range of Amb.+15°C to 250°C. The advanced model offers enhanced functionality such as programmable temperature ramping, a real-time graphical display, and RS-232/USB communication ports for data logging and external control. With PID temperature control, a durable corrosion-resistant chamber, and a user-friendly interface, the ON4 series delivers dependable performance for precise, disturbance-free heating applications. Inner chamber: W 19.69 x D 17.56 x H 25.95 inches.
Catalog No. NC3965881 Supplier Jeio Tech Inc Supplier No. AAH111136U
Add to Cart
Add to Cart
Jeio Tech ON4-15V Natural Convection (Gravity) Oven is engineered for gentle, reliable thermal processing with uniform temperature distribution, ideal for drying heat-sensitive materials, powders, and glassware without airflow disturbance. Utilizing natural gravity convection, the PID control system delivers excellent stability (±0.5°C fluctuation) and uniformity (±3.5°C variation) across a temperature range of Amb.+15°C to 250°C. The advanced model offers enhanced functionality such as programmable temperature ramping, a real-time graphical display, and RS-232/USB communication ports for data logging and external control. With PID temperature control, a durable corrosion-resistant chamber, and a user-friendly interface, the ON4 series delivers dependable performance for precise, disturbance-free heating applications. Inner chamber: W 19.69 x D 17.56 x H 25.95 inches.