Learn More
Jeio Tech Inc Jeio Tech™ RS-2 Vertical Funnel Shaker (Incl. 4ea holders) (100-240V, 50/60Hz), Liquid-Liquid Extraction, Adjustable Tilt, Multi-Funnel Capacity, Quiet Motor, Hands-Free Mixing
SDP
Catalog No. NC3965949
Encompass_Preferred
Jeio Tech RS-2 Vertical Funnel Shaker, Liquid-Liquid Extraction with Adjustable Tilt Platform delivers efficient, reproducible mixing for laboratory workflows. Its vertical shaking motion with 40 mm stroke enhances separation performance and consistency, outperforming manual methods. The system supports up to 2L separating funnels and includes 4 holders (expandable to 10), enabling high-throughput operation. An adjustable platform tilts up to 20° to optimize mixing based on solvent properties. Designed for safety and ease of use, it features hands-free operation, a built-in timer, speed deviation alarm, and overcurrent protection. With a quiet motor, durable construction, and user-friendly controls, the RS-2 is ideal for environmental, chemical, and pharmaceutical labs requiring precise extraction.
Catalog No. NC3965949 Supplier Jeio Tech Inc Supplier No. AAH32023U
Jeio Tech RS-2 Vertical Funnel Shaker, Liquid-Liquid Extraction with Adjustable Tilt Platform delivers efficient, reproducible mixing for laboratory workflows. Its vertical shaking motion with 40 mm stroke enhances separation performance and consistency, outperforming manual methods. The system supports up to 2L separating funnels and includes 4 holders (expandable to 10), enabling high-throughput operation. An adjustable platform tilts up to 20° to optimize mixing based on solvent properties. Designed for safety and ease of use, it features hands-free operation, a built-in timer, speed deviation alarm, and overcurrent protection. With a quiet motor, durable construction, and user-friendly controls, the RS-2 is ideal for environmental, chemical, and pharmaceutical labs requiring precise extraction.