Learn More
Jeio Tech Inc Jeio Tech™ ST-105G, 105L Vertical Autoclave (230V, 50/60Hz, NEMA 6-30 Plug), Programmable Sterilizer with Stable Temperature Control, Safety Protection, Space-Saving, Top-Loading Design
SDP
Catalog No. NC3965837
Encompass_Preferred
Jeio Tech ST-105G Autoclave, Vertical Top-Loading Sterilizer delivers reliable, consistent sterilization for glassware, instruments, and media. Designed for ease of use, it features stable temperature control and a spacious chamber for efficient loading and cleaning. The intuitive LED/VFD control panel supports programmable sterilization cycles for diverse applications. Built-in safety systems include overpressure, overtemperature, overcurrent protection, and low water-level alarm for secure operation. Optional accessories such as a digital pressure sensor, data recorder, and printer enhance data management, making the ST-G a dependable and user-friendly solution for laboratory and industrial sterilization needs. Inner chamber: Diameter 15.75" x H 35.75".
Catalog No. NC3965837 Supplier Jeio Tech Inc Supplier No. AAHL1045K
Jeio Tech ST-105G Autoclave, Vertical Top-Loading Sterilizer delivers reliable, consistent sterilization for glassware, instruments, and media. Designed for ease of use, it features stable temperature control and a spacious chamber for efficient loading and cleaning. The intuitive LED/VFD control panel supports programmable sterilization cycles for diverse applications. Built-in safety systems include overpressure, overtemperature, overcurrent protection, and low water-level alarm for secure operation. Optional accessories such as a digital pressure sensor, data recorder, and printer enhance data management, making the ST-G a dependable and user-friendly solution for laboratory and industrial sterilization needs. Inner chamber: Diameter 15.75" x H 35.75".