Learn More
ATCC JeKo-1; Mantle Cell Lymphoma; Human (Homo sapiens)
Catalog No. 502383815
Encompass
The JeKo-1 mantle cell lymphoma (MCL) cell line was isolated from the peripheral blood mononuclear cells of a 78-year-old female with a large cell variant of MCL showing leukemic conversion. The JeKo-1 cells are highly tumorigenic in SCID mice and has applications for immunology research.
Catalog No. 50-238-3815 Supplier ATCC Supplier No. CRL3006
The JeKo-1 mantle cell lymphoma (MCL) cell line was isolated from the peripheral blood mononuclear cells of a 78-year-old female with a large cell variant of MCL showing leukemic conversion. The JeKo-1 cells are highly tumorigenic in SCID mice and has applications for immunology research.