Learn More
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Lyophilized from buffer salt primarily Sodium EDTA, pH 7.5 |
| Name | Jellyfish aequorin native Protein from |
| Quantity | 25 μg |
| Storage Requirements | Store at −20°C on dry atmosphere.After reconstitution, aliquot and store at 2−8°C. Reconstitution: Reconstitute with calcium free water or other appropriate buffer which is calcium free. |
| Regulatory Status | RUO |
| Common Name | AEQUORIN |
| Species | Jellyfish |
| Protein Tag | None |
| Expression System | Native protein purified from jellyfish |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.