Learn More
Enterprise Technology Solutions Jelmar LLC Cleaner,f/Calcium&Lime&Rust,Bleach&AmoniaFree,5Gal,White
SDP
Catalog No. 5000830836
Encompass_Preferred
CLR PRO Calcium, Lime & Rust Remover is the fast and effective way to remove calcium, lime and rust deposits. This industrial-strength, multi-purpose cleaner is designed to quickly remove calcium, lime and hard-water deposits, soap scum, discoloration and dirt. Use on kitchen or bathroom related surfaces, like plastic, ceramic tile, glass, stainless steel, faucet and shower fixtures, shower doors, fiberglass, toilet bowls and sinks. Cleaner is part of the EPA's Safer Choice Program, recognizing the product as a safer alternative to traditional chemicals, and contains no phosphates, ammonia or bleach. Calcium, lime and rust remover is also NSF registered and is approved for use in food processing, food storage, and food service facilities.
Catalog No. 50-008-30836 Supplier Enterprise Technology Solutions Supplier No. JELFMCLR5PRO
Add to Cart
Add to Cart
CLR PRO Calcium, Lime & Rust Remover is the fast and effective way to remove calcium, lime and rust deposits. This industrial-strength, multi-purpose cleaner is designed to quickly remove calcium, lime and hard-water deposits, soap scum, discoloration and dirt. Use on kitchen or bathroom related surfaces, like plastic, ceramic tile, glass, stainless steel, faucet and shower fixtures, shower doors, fiberglass, toilet bowls and sinks. Cleaner is part of the EPA's Safer Choice Program, recognizing the product as a safer alternative to traditional chemicals, and contains no phosphates, ammonia or bleach. Calcium, lime and rust remover is also NSF registered and is approved for use in food processing, food storage, and food service facilities.