Learn More
Enterprise Technology Solutions Jelmar LLC Cleaner,TarnishRemover,f/Copper&SterlingSilv,1Gal,4/CT,WE
SDP
Catalog No. 5000830852
Encompass_Preferred
Tarn-X PRO Tarnish Remover is the leading tarnish remover for cleaning and brightening copper and sterling silver. Non-flammable quick dip, non-scrubbing formula saves time and effort with no buffing required. Remover also rinses clean with no residue. For best results, spot test in an inconspicuous area first.
Catalog No. 50-008-30852 Supplier Enterprise Technology Solutions Supplier No. JELFMTX1284PRO
Tarn-X PRO Tarnish Remover is the leading tarnish remover for cleaning and brightening copper and sterling silver. Non-flammable quick dip, non-scrubbing formula saves time and effort with no buffing required. Remover also rinses clean with no residue. For best results, spot test in an inconspicuous area first.