Learn More
Newcomer Supply Jenner Stock Stain, 500 ml
SDP
Catalog No. NC9527282
Encompass
Use: May-Grunwald mod. Giemsa Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC9527282 Supplier Newcomer Supply Supplier No. 1210A
Use: May-Grunwald mod. Giemsa Stain. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.