Learn More
Bioworld Jensen's Medium, 50 L
Catalog No. 50254632
Encompass_Preferred
For nodulation studiesSpecifications:Powder mediumContains (g/L): 1g Calcium phosphate, 0.2g Potassium phosphate, 0.2g Magnesium sulphate, 0.2g Sodium chloride and 0.1g Ferric chloride. Final pH: 7.0 ±0.2 @ 25°CProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedDirection to use: Suspend 1.7g medium with 12g agar in one liter distilled water. Heat to dissolve completly and autoclave to sterilize at 121°C for 15 minutes.
Catalog No. 50-254-632 Supplier Bioworld Supplier No. 306300394
For nodulation studiesSpecifications:Powder mediumContains (g/L): 1g Calcium phosphate, 0.2g Potassium phosphate, 0.2g Magnesium sulphate, 0.2g Sodium chloride and 0.1g Ferric chloride. Final pH: 7.0 ±0.2 @ 25°CProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedDirection to use: Suspend 1.7g medium with 12g agar in one liter distilled water. Heat to dissolve completly and autoclave to sterilize at 121°C for 15 minutes.