Learn More
Cole-Parmer Jenway 3510 Standard Digital pH Meter Only, ATC; 120 V
Supplier:  Cole-Parmer 3545903
Encompass_Preferred
Benchtop pH meters feature up to three decimal place resolution, support up to three pH calibration points, and offer manual and automatic buffer recognition for DIN, JIS, and NIST buffers. A large LCD displays status and results of pH and temperature readings. Data is easily exported to a computer or printer via RS-232 connection.
Catalog No. NC1239144