Learn More
Electron Microscopy Sciences JEOL 2-Position Single Tilt Holder
SDP
Catalog No. 501930544
Encompass_Preferred
A single tilt multi-grid holder. Will accommodate 6 grids with an easy click stop reproducible movement between each specimen. It is ideal for scanning through serial sections or for comparing against a standard. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own mahogany box.
Catalog No. 50-193-0544 Supplier Electron Microscopy Sciences Supplier No. EMS027
Add to Cart
Add to Cart
Request a Quote
A single tilt multi-grid holder. Will accommodate 6 grids with an easy click stop reproducible movement between each specimen. It is ideal for scanning through serial sections or for comparing against a standard. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own mahogany box.