Learn More
Electron Microscopy Sciences JEOL 2 Position Single Tilt Holder
SDP
Catalog No. 501930521
Encompass_Preferred
A single tilt two-grid sample holder. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own mahogany box.
Catalog No. 50-193-0521 Supplier Electron Microscopy Sciences Supplier No. EMS017
Add to Cart
Add to Cart
Request a Quote
A single tilt two-grid sample holder. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own mahogany box.