Learn More
Electron Microscopy Sciences JEOL 3 Position Grid Insert
SDP
Catalog No. 501930546
Encompass_Preferred
A three position insert which will fit standard JEOL 2 position holders as a direct replacement. Specimens are held in place with our push fit, easy to use circlips. The circlip extraction tool, Cat. #EMS016 is needed to install and remove circlips.
Catalog No. 50-193-0546 Supplier Electron Microscopy Sciences Supplier No. EMS056
A three position insert which will fit standard JEOL 2 position holders as a direct replacement. Specimens are held in place with our push fit, easy to use circlips. The circlip extraction tool, Cat. #EMS016 is needed to install and remove circlips.