Learn More
Electron Microscopy Sciences Applied Physics CeB6 Cathode for JEOL
SDP
Catalog No. 50949902
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: CEBIX (CeB6)
Angle: 90°
Tips: 16 mm
Catalog No. 50-949-902 Supplier Electron Microscopy Sciences Supplier No. 80927
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: CEBIX (CeB6)
Angle: 90°
Tips: 16 mm