Learn More
MSC Jergens Center-Pull Hoist Ring, 11000 kg Capacity, M36 x 4.0 Thread, Black Oxide Alloy Steel
Catalog No. 5000529918
Encompass
This center-pull hoist ring is engineered for high-capacity lifting applications, featuring a robust design and durable construction. It offers full 360° swivel and 180° pivot action, ensuring versatile and secure load handling.
  • Rated at 5:1 strength factor
  • Constructed from alloy steel with a black oxide finish
  • U-bar, bolt, pins, base, and bushing are magnetic particle inspected
  • Proof tested to 200% of rated load capacity
  • Certificate of proof test provided
  • Conforms to ASME B30.26 and MIL-STD-209K
  • Operating temperature range from -28°C to 204°C
Catalog No. 50-005-29918 Supplier MSC Supplier No. 04555603
This center-pull hoist ring is engineered for high-capacity lifting applications, featuring a robust design and durable construction. It offers full 360° swivel and 180° pivot action, ensuring versatile and secure load handling.
  • Rated at 5:1 strength factor
  • Constructed from alloy steel with a black oxide finish
  • U-bar, bolt, pins, base, and bushing are magnetic particle inspected
  • Proof tested to 200% of rated load capacity
  • Certificate of proof test provided
  • Conforms to ASME B30.26 and MIL-STD-209K
  • Operating temperature range from -28°C to 204°C