Learn More
MSC Jergens Center-Pull Hoist Ring, 30000 lb Capacity, 2-4 1/2 Thread, Alloy Steel
Catalog No. 5000535662
Encompass
This center-pull hoist ring is engineered for demanding lifting tasks, offering a robust load capacity and a secure bolt-on mount. It is constructed from durable alloy steel with a black oxide finish for enhanced longevity and reliable performance.
  • Provides a load capacity of 30,000 lb.
  • Features a 2-4 1/2 thread size.
  • Requires an installation torque of 1100 ft-lb.
  • Made from domestically-produced, certified alloy steel.
  • All critical components are magnetic particle inspected.
  • Conforms to ASME B30.26 and MIL-STD-209K standards.
Catalog No. 50-005-35662 Supplier MSC Supplier No. 82501412
This center-pull hoist ring is engineered for demanding lifting tasks, offering a robust load capacity and a secure bolt-on mount. It is constructed from durable alloy steel with a black oxide finish for enhanced longevity and reliable performance.
  • Provides a load capacity of 30,000 lb.
  • Features a 2-4 1/2 thread size.
  • Requires an installation torque of 1100 ft-lb.
  • Made from domestically-produced, certified alloy steel.
  • All critical components are magnetic particle inspected.
  • Conforms to ASME B30.26 and MIL-STD-209K standards.