Learn More
MSC Jergens Hoist Ring, Safety Engineered Center Pull, 7000 lb Capacity, 3/4-10 Thread
Catalog No. 5000527876
Encompass
The product is a Safety Engineered Center Pull Hoist Ring with a 7000 lb capacity, featuring a 3/4-10 thread and constructed from alloy steel. This hoist ring is designed for secure lifting applications.
  • Safety engineered center pull type
  • 7000 lb load capacity
  • 3/4-10 thread size
  • Alloy steel construction
  • 1.00 in ring thickness
  • 2.31 in ring internal width
  • 5.14 in ring external width
Catalog No. 50-005-27876 Supplier MSC Supplier No. 36926525
The product is a Safety Engineered Center Pull Hoist Ring with a 7000 lb capacity, featuring a 3/4-10 thread and constructed from alloy steel. This hoist ring is designed for secure lifting applications.
  • Safety engineered center pull type
  • 7000 lb load capacity
  • 3/4-10 thread size
  • Alloy steel construction
  • 1.00 in ring thickness
  • 2.31 in ring internal width
  • 5.14 in ring external width