Learn More
MSC Jergens Hoist Ring, Safety Engineered Center Pull, Bolt-On, 10000 lb Capacity, Alloy Steel
Catalog No. 5000527591
Encompass
Jergens Safety Engineered Center Pull Hoist Rings are designed for secure lifting applications, featuring a bolt-on mount and a robust 10000 lb load capacity. Constructed from durable alloy steel with a black oxide finish, this hoist ring ensures reliability and safety in various industrial environments.
  • Safety engineered center pull design
  • Bolt-on mount type
  • 10000 lb load capacity
  • Alloy steel material
  • Black oxide finish
  • CE standards compliant
  • Material grade 4140
  • Thread size 1-8
  • Torque 230 FT/LB
  • Single mounting hole
Catalog No. 50-005-27591 Supplier MSC Supplier No. 82501370
Jergens Safety Engineered Center Pull Hoist Rings are designed for secure lifting applications, featuring a bolt-on mount and a robust 10000 lb load capacity. Constructed from durable alloy steel with a black oxide finish, this hoist ring ensures reliability and safety in various industrial environments.
  • Safety engineered center pull design
  • Bolt-on mount type
  • 10000 lb load capacity
  • Alloy steel material
  • Black oxide finish
  • CE standards compliant
  • Material grade 4140
  • Thread size 1-8
  • Torque 230 FT/LB
  • Single mounting hole