Learn More
MSC Jergens Safety Engineered Center Pull Hoist Ring, Bolt-On, 24000 lb, 1-1/2-6
Catalog No. 5000532117
Encompass
This safety engineered center pull hoist ring is designed for secure lifting applications. It features a bolt-on mounting type and a 24,000 lb capacity. The hoist ring is constructed from black oxide alloy steel and has a 1-1/2-6 thread size.
  • Safety engineered center pull design
  • Work load limit of 24,000 lb
  • 1-1/2-6 thread size
  • Constructed from alloy steel
  • Black oxide finish
  • Bolt-on mount type
  • CE standard compliant
  • Single mounting hole
Catalog No. 50-005-32117 Supplier MSC Supplier No. 82501404
This safety engineered center pull hoist ring is designed for secure lifting applications. It features a bolt-on mounting type and a 24,000 lb capacity. The hoist ring is constructed from black oxide alloy steel and has a 1-1/2-6 thread size.
  • Safety engineered center pull design
  • Work load limit of 24,000 lb
  • 1-1/2-6 thread size
  • Constructed from alloy steel
  • Black oxide finish
  • Bolt-on mount type
  • CE standard compliant
  • Single mounting hole