Learn More
MSC Jergens Safety Engineered Center Pull Hoist Ring, Bolt-On, 7000 kg Capacity, M30 x 3.5 Thread
Catalog No. 5000529459
Encompass
This safety engineered center pull hoist ring is designed for bolt-on applications, offering a robust 7000 kg capacity. It features an M30 x 3.5 thread and is constructed from black oxide alloy steel for durability and performance.
  • Safety engineered center pull hoist ring type
  • Bolt-on mount type
  • Black oxide finish
  • 4140 material grade
  • Single mounting hole
  • CE standards compliant
  • 7000 kg load capacity
  • 44.50 mm radius
  • M30 x 3.5 thread size
  • 221.70 mm ring height
  • Alloy steel material
  • 31.70 mm ring thickness
  • 165.10 mm ring external width
  • 81.00 mm ring internal width
  • 31.70 mm diameter
Catalog No. 50-005-29459 Supplier MSC Supplier No. 04555587
This safety engineered center pull hoist ring is designed for bolt-on applications, offering a robust 7000 kg capacity. It features an M30 x 3.5 thread and is constructed from black oxide alloy steel for durability and performance.
  • Safety engineered center pull hoist ring type
  • Bolt-on mount type
  • Black oxide finish
  • 4140 material grade
  • Single mounting hole
  • CE standards compliant
  • 7000 kg load capacity
  • 44.50 mm radius
  • M30 x 3.5 thread size
  • 221.70 mm ring height
  • Alloy steel material
  • 31.70 mm ring thickness
  • 165.10 mm ring external width
  • 81.00 mm ring internal width
  • 31.70 mm diameter