Learn More
MSC Jet Hydraulic Bottle Jack, 8 Ton Capacity, 7-7/8-15-1/2 in Height
Catalog No. 5000528376
Encompass
This 8-Ton Hydraulic Bottle Jack is a robust and versatile tool for demanding lifting, pushing, spreading, bending, pressing, or straightening applications. Its industrial-duty construction ensures reliable performance in extreme climates, with precision machined cylinders providing leak-free operation.
  • Complies with ANSI/ASME B30.1 and PALD-1 standard
  • Heavy-duty, electrically welded base for stability
  • Internal by-pass mechanism protects hydraulic system
  • Industrial-duty construction for extreme climates
  • Versatile for lifting, pushing, spreading, bending, pressing, or straightening
  • Operates from -4°F to 158°F
  • Precision machined, polished cylinders provide leak-free performance
  • Includes pump handle
  • Supports upright, horizontal, or angled operating positions
Catalog No. 50-005-28376 Supplier MSC Supplier No. 37614302
This 8-Ton Hydraulic Bottle Jack is a robust and versatile tool for demanding lifting, pushing, spreading, bending, pressing, or straightening applications. Its industrial-duty construction ensures reliable performance in extreme climates, with precision machined cylinders providing leak-free operation.
  • Complies with ANSI/ASME B30.1 and PALD-1 standard
  • Heavy-duty, electrically welded base for stability
  • Internal by-pass mechanism protects hydraulic system
  • Industrial-duty construction for extreme climates
  • Versatile for lifting, pushing, spreading, bending, pressing, or straightening
  • Operates from -4°F to 158°F
  • Precision machined, polished cylinders provide leak-free performance
  • Includes pump handle
  • Supports upright, horizontal, or angled operating positions