Learn More
MSC Jet Manual Puller Hoist, 1000 lb Capacity, 15 ft Max Lift
Catalog No. 5000514315
Encompass
This manual puller hoist from the JLP-A Series is designed for light to medium-duty industrial applications, emphasizing safety, reliability, and durability. It features a 1000 lb capacity and a 15 ft maximum lift.
  • Weston Style, double pawl brake design for enhanced load security.
  • Industrial-rated alloy steel hooks rotate 360 degrees and indicate overload.
  • Brake wear indicator provides a visual cue for disc replacement.
  • Cast steel safety latches with Allen screws and lock nuts.
  • Grade 80 black oxide coating and impact-resistant all-steel construction for durability.
  • Needle bearing design improves efficiency and smooth operation.
  • Compact and lightweight design ideal for low headroom and confined areas.
  • Complies with ANSI / ASME B30.21 and HST-3 standards.
  • Load tested to 125% of capacity with a certificate.
Catalog No. 50-005-14315 Supplier MSC Supplier No. 42342444
This manual puller hoist from the JLP-A Series is designed for light to medium-duty industrial applications, emphasizing safety, reliability, and durability. It features a 1000 lb capacity and a 15 ft maximum lift.
  • Weston Style, double pawl brake design for enhanced load security.
  • Industrial-rated alloy steel hooks rotate 360 degrees and indicate overload.
  • Brake wear indicator provides a visual cue for disc replacement.
  • Cast steel safety latches with Allen screws and lock nuts.
  • Grade 80 black oxide coating and impact-resistant all-steel construction for durability.
  • Needle bearing design improves efficiency and smooth operation.
  • Compact and lightweight design ideal for low headroom and confined areas.
  • Complies with ANSI / ASME B30.21 and HST-3 standards.
  • Load tested to 125% of capacity with a certificate.