Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JetFlex™ Genomic DNA Purification Kit

Catalog No. p-7181373
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
500 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A30701 500 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A30701 Supplier Invitrogen™ Supplier No. A30701
Only null left
Add to Cart
Edge
Add to Cart

The JetFlex™ Genomic DNA Purification Kit is designed for rapid and efficient purification of genomic DNA (gDNA) from blood, body fluids (amniotic fluid, saliva, sperm, lymph), mammalian cells, tissues, plant tissues, bacteria, and yeast.

Single-tube technology allows for isolation of high quality, high molecular weight, RNA-free gDNA without the use of spin columns of a vacuum manifold.

Key benefits include:

  • Large volume processing of 2 mL of whole blood or other body fluid
  • Capable of processing:
    • 2 x 107 mammalian cells
    • 10 mg of tissue
    • 500 μL bacterial culture
    • 2 x 108 cells/mL yeast culture
  • Simple protocol that does not require columns or vacuum manifolds
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 x 165 mL RBC Lysis Buffer, Store at Room Temperature
• 1 x 165 mL Cell Lysis Buffer, Store at Room Temperature
• 1 x 83 mL PPT Buffer, Store at Room Temperature
• 1 x 30 mL Pellet Compactor, Store at Room Temperature
• 1 x 110 mL TE Buffer, Store (at Room Temperature
• 10 x 21 mg Proteinase K (lyophilized powder), Store 2°C to 8°C
• 5 x 1.1 mL RNase A, Store 2°C to 8°C
Format Spin Column
Isolation Technology Tube Centrifugation
Sample Type Blood, Buccal Samples, Cells, Liquid Samples (e.g. Serum)
For Use With (Application) Real-Time PCR, PCR
High-throughput Compatibility Not High-throughput Compatible
No. of Reactions 500 Preps
Product Line JetFlex
Product Type Genomic DNA Purification Kit
Quantity 500 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.