Learn More
Genesee Scientific Corporation jetPEI HTS DNA Transfection Reagent, Low Toxicity, High Throughput Screening Compatible, 5 °C ± 3°C
SDP
Catalog No. NC1937527
Encompass
jetPEI HTS DNA Transfection Reagent is a linear polyethylenimine derivative designed for efficient gene delivery in High Throughput Screening (HTS) applications. It features low toxicity, and compatibility with serum and antibiotics, enhancing experimental flexibility. Its exceptional reproducibility ensures reliable results across experiments, making it an ideal choice for both automated and manual workflows.
Catalog No. NC1937527 Supplier Genesee Scientific Corporation Supplier No. 55102N
jetPEI HTS DNA Transfection Reagent is a linear polyethylenimine derivative designed for efficient gene delivery in High Throughput Screening (HTS) applications. It features low toxicity, and compatibility with serum and antibiotics, enhancing experimental flexibility. Its exceptional reproducibility ensures reliable results across experiments, making it an ideal choice for both automated and manual workflows.