Learn More
Andwin Scientific JETPRIME DNA/SIRNA RGNT 5ML
SDP
Catalog No. NC0291070
Encompass_Preferred
The jetPRIME DNA and siRNA Transfection Reagent is made to guarantee high DNA transfection efficiency and excellent gene silencing. This product is very cost-effective as it uses less reagent and DNA and provides more gentle transfections in comparison to other competitors. It can be used with no difficulty and with serum and antibiotics. This can produce 15 reactions in 6-well plates and has a buffer volume of 5 mL. It is sold individually.
Catalog No. NC0291070 Supplier Andwin Scientific Supplier No. 89129920
The jetPRIME DNA and siRNA Transfection Reagent is made to guarantee high DNA transfection efficiency and excellent gene silencing. This product is very cost-effective as it uses less reagent and DNA and provides more gentle transfections in comparison to other competitors. It can be used with no difficulty and with serum and antibiotics. This can produce 15 reactions in 6-well plates and has a buffer volume of 5 mL. It is sold individually.