Learn More
Andwin Scientific JETPRIME SIZE: 5 X 1.5ML + BUF
SDP
Catalog No. NC1948794
Encompass_Preferred
Jetprime Size: 5 X 1.5Ml + Buffer^ Jetprime Is A NewPowerful & Polyvalent Transfection Reagent DesignedFor Efficient Delivery Of Dna And Sirna Across ManyAdherent Cell Types. Jetprime Provides HigherTransfection Efficiency, Uses Less Reagent, Is CostEffective, And Delivers Gentle Transfections.
Catalog No. NC1948794 Supplier Andwin Scientific Supplier No. 89129926
Jetprime Size: 5 X 1.5Ml + Buffer^ Jetprime Is A NewPowerful & Polyvalent Transfection Reagent DesignedFor Efficient Delivery Of Dna And Sirna Across ManyAdherent Cell Types. Jetprime Provides HigherTransfection Efficiency, Uses Less Reagent, Is CostEffective, And Delivers Gentle Transfections.