Learn More
Sklar Instruments Forceps, Merit Jeweler Style Forceps, Sklar 4-1/2 in., Sharp, Straight, Smooth, #5, Stainless Steel, EA.
SDP
Catalog No. 501126821 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Merit Jeweler Style Forceps, Sklar, 4-1/2 in., Sharp, Straight, Smooth, #5, Stainless Steel, EA. Merit Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/4 inches. It is also known as the #5 style jeweler forceps.
Catalog No. 50-112-6821 Supplier Sklar Instruments Supplier No. 98817
Forceps, Merit Jeweler Style Forceps, Sklar, 4-1/2 in., Sharp, Straight, Smooth, #5, Stainless Steel, EA. Merit Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit instruments deliver an economical solution for physicians and practices that require quality, durable instruments. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/4 inches. It is also known as the #5 style jeweler forceps.