Learn More
Sklar Instruments Forceps, Jeweler Forceps, Sklar 4-1/2 in., Sharp, Straight, Smooth, #1, EA.
SDP
Catalog No. 501119025 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Jeweler Forceps, Sklar, 4-1/2 in., Sharp, Straight, Smooth, #1, EA. Sklar Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/2 inches. It is also known as the #1 style jeweler forceps. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-9025 Supplier Sklar Instruments Supplier No. 667438
Forceps, Jeweler Forceps, Sklar, 4-1/2 in., Sharp, Straight, Smooth, #1, EA. Sklar Jeweler Style Forceps are grasping forceps used in microsurgical procedures. They have fine or micro-fine tips that can handle extremely fine tissues and materials that standard instruments may damage. They are multipurpose thumb forceps with flat handles available with various tip styles. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/2 inches. It is also known as the #1 style jeweler forceps. Lifetime warranty against all manufacturer defects.