Learn More
Electron Microscopy Sciences Jewelers Saw Blade Size 1/0
SDP
Supplier:  Electron Microscopy Sciences 7202001
Encompass_Preferred
Swiss-made jewlers saw with a fully adjustable frame that permits the tightening of the blade as well as using broken blades. Each blade has an overall length of 5¼" (133 mm).
Thickness: 0.28 mm
Width: 0.58 mm
Teeth: 20 teeth per cm
Catalog No. 50-949-097