Learn More
Electron Microscopy Sciences Jewelers Saw Blade Size 4/0
SDP
Catalog No. 50949100
Encompass_Preferred
Swiss-made jewlers saw with a fully adjustable frame that permits the tightening of the blade as well as using broken blades. Each blade has an overall length of 5¼" (133 mm).
Thickness: 0.23 mm
Width: 0.43 mm
Teeth: 25 teeth per cm
Catalog No. 50-949-100 Supplier Electron Microscopy Sciences Supplier No. 7202004
Swiss-made jewlers saw with a fully adjustable frame that permits the tightening of the blade as well as using broken blades. Each blade has an overall length of 5¼" (133 mm).
Thickness: 0.23 mm
Width: 0.43 mm
Teeth: 25 teeth per cm