Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Jewett™ High-Performance Lab Freezers

Catalog No. JLF430A Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (English):
4.7 cu. ft.
Plug Type:
NEMA 5-15 P
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Plug Type Voltage
JLF430A 4.7 cu. ft. NEMA 5-15 P 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. JLF430A Supplier Thermo Scientific™ Supplier No. JLF430A

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Store reagents, pharmaceuticals, biologicals and other commonly used laboratory materials.

Store reagents, pharmaceuticals, biologicals and other commonly used laboratory materials in Thermo Scientific™ Jewett™ High-Performance Lab Freezers. Designed for medical and scientific applications, these high-performance freezers feature microprocessor control, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors selected for industrial, clinical and scientific use.

Microprocessor Control

  • Microprocessor control system with audio and visual alarms
  • Bright digital temperature displays
  • Alarm silence, ringback and automatic reset functions
  • Setpoint security with key-operated, triple-position master switch that controls power to all systems
  • Audible/visual alarms for high/low temperature and power failure
  • Battery backup and remote alarm connections
  • Temperature is factory-preset at -30°C.†

Construction

  • Rugged, long-life cabinets with smooth, scratch-resistant interiors‡ and exteriors that are easy to clean
  • High-density, CFC-free insulation
  • Solid self-closing doors with 90° stay-open feature, keyed lock for added sample security
  • Closed-cell gasket provides a positive door seal to maintain a constant interior temperature
  • Directed airflow for fast temperature recovery after door openings
  • Smooth, ergonomic full-length door handle
  • Painted open-wire shelves are adjustable in 1″ (2.5cm) increments
  • 1″ (2.5cm) insulated access port standard
  • Set of four 2″ (5.2cm) casters included, two locking and two regular

Refrigeration System

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Automatic defrost maintains optimum cooling

Ordering Information:

Options include freestanding low-voltage booster and built-in seven-day temperature chart recorder. Contact your Customer Service Representative for more information. V and W models not available in North America.

Note:

† Performance may be affected by ambient temperature and humidity conditions, sample loading preferences, and frequency of door openings.

‡ Specified interior volumes are nominal, and usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions..

TRUSTED_SUSTAINABILITY

Specifications

Type High-Performance Freezer
Capacity (English) 4.7 cu. ft.
Capacity (Metric) 133 L
Temperature Range -30°C
Door Style Solid
No. of Doors 1
Insulation CFC-free
Shelves 3
Certifications/Compliance CE, UL
Style Undercounter
Control Type Microprocessor
Warranty Warranty varies by country, please contact us for details
Defrost Automatic
Regulatory Approvals Thermo Fisher Scientific is committed to meeting all compliance obligations under the California Cooling Act. Certain products are still transitioning into using acceptable appliance foams and/or refrigerants. This product is currently not available for sale into the state of California.
Dimensions (D x W x H) Interior 20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8 cm)
Dimensions (L x W x H) Exterior 26 x 24 x 33.4 in. (66 x 61 x 85 cm)
Depth (English) Interior 20.5 in.
Depth (Metric) Interior 52.1 cm
Height (English) Exterior 33.4 in.
Height (English) Interior 20 in.
Height (Metric) Exterior 85 cm
Height (Metric) Interior 50.8 cm
Length (English) Exterior 26 in.
Length (Metric) Exterior 66 cm
Width (English) Exterior 24 in.
Width (English) Interior 20 in.
Width (Metric) Exterior 61 cm
Width (Metric) Interior 50.8 cm
Monitoring Options Alarm, Recorder
Amperage 8 A
Breaker 15A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Compressor Heavy-duty, Industrial-grade, Hermetic
Refrigerant CFC-free
Shipping Weight (English) 220 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.