Learn More
Medchemexpress LLC HY-117282 10mg Medchemexpress, JG-98 CAS:1456551-16-8 Purity:>98%
SDP
Catalog No. 501969463
Encompass_Preferred
Medchemexpress, HY-117282 10mg JG-98 CAS:1456551-16-8 JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9463 Supplier Medchemexpress LLC Supplier No. HY11728210MG
Medchemexpress, HY-117282 10mg JG-98 CAS:1456551-16-8 JG-98, an allosteric heat shock protein 70 (Hsp70) inhibitor, which binds tightly to a conserved site on Hsp70 and disrupts the Hsp70-Bag3 interaction. JG-98 shows anti-cancer activities affecting both cancer cells and tumor-associated macrophages[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.