Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH-II-127 | 1700693-08-8 | 98.0% | 416.86 | 1 ML
SDP

Catalog No. 5000391110
Encompass_Preferred

JH-II-127 is an orally active, highly potent, selective, and brain-permeable LRRK2 inhibitor. It has IC50s of 6, 2, and 48 nM for wild-type LRRK2, LRRK2-G2019S, and mutant LRRK2-A2016T, respectively. This compound inhibits Ser935 phosphorylation in all tissues of mice, including the brain. JH-II-127 can be utilized in the study of Parkinson's syndrome.

  • CAS No.: 1700693-08-8
  • Molecular weight: 416.86
  • Purity: 98.0%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C19H21ClN6O3
  • Solubility in vitro: DMSO: 110 mg/mL (263.88 mM)
  • Solubility in vivo dissolution protocols available
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 2 years, -20°C for 1 year

Catalog No. 50-003-91110 Supplier Medchemexpress LLC Supplier No. HY1693610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH-II-127 is an orally active, highly potent, selective, and brain-permeable LRRK2 inhibitor. It has IC50s of 6, 2, and 48 nM for wild-type LRRK2, LRRK2-G2019S, and mutant LRRK2-A2016T, respectively. This compound inhibits Ser935 phosphorylation in all tissues of mice, including the brain. JH-II-127 can be utilized in the study of Parkinson's syndrome.

  • CAS No.: 1700693-08-8
  • Molecular weight: 416.86
  • Purity: 98.0%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Formula: C19H21ClN6O3
  • Solubility in vitro: DMSO: 110 mg/mL (263.88 mM)
  • Solubility in vivo dissolution protocols available
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.