Learn More
Medchemexpress LLC Jh-re-06 | 1361227-90-8 | 99.9% | 468.72 | C20H16Cl3N3O4 | 10MG
SDP
Catalog No. 5000239228
Encompass_Preferred
JH-RE-06 is a small-molecule REV1-POLζ interface inhibitor used in research to disrupt mutagenic translesion synthesis and sensitize cells to chemotherapy. It is provided as a solid (white to yellow) with high purity, molecular formula C20H16Cl3N3O4, and molecular weight 468.72 g/mol. Recommended storage conditions are powder: -20°C (stable up to 3 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Potent REV1-POLζ interface inhibitor for translesion synthesis studies.
  • High purity (99.9%) suitable for analytical and biological assays.
  • Solid form with documented storage conditions to preserve stability.
  • Well characterized with known molecular formula and molecular weight.
  • Supporting documentation available including data sheet, COA, and safety data sheet.
Catalog No. 50-002-39228 Supplier Medchemexpress LLC Supplier No. HY12621410MG
JH-RE-06 is a small-molecule REV1-POLζ interface inhibitor used in research to disrupt mutagenic translesion synthesis and sensitize cells to chemotherapy. It is provided as a solid (white to yellow) with high purity, molecular formula C20H16Cl3N3O4, and molecular weight 468.72 g/mol. Recommended storage conditions are powder: -20°C (stable up to 3 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).
  • Potent REV1-POLζ interface inhibitor for translesion synthesis studies.
  • High purity (99.9%) suitable for analytical and biological assays.
  • Solid form with documented storage conditions to preserve stability.
  • Well characterized with known molecular formula and molecular weight.
  • Supporting documentation available including data sheet, COA, and safety data sheet.