Learn More
Medchemexpress LLC JH-RE-06 | 1361227-90-8 | 99.9% | 468.72 | C20H16Cl3N3O4 | 5MG
SDP
Catalog No. 5000234980
Encompass_Preferred
JH-RE-06 is a small-molecule REV1-REV7 interface inhibitor that disrupts mutagenic translesion synthesis (TLS) by preventing recruitment of POLζ, sensitizing cells to DNA-damaging chemotherapy. It is characterized in cellular and in vivo models for enhancing cisplatin efficacy and reducing therapy-induced mutagenesis; intended for research use only.
  • Targets REV1-REV7 interface to inhibit TLS.
  • Sensitizes tumor cells to DNA-damaging agents.
  • Demonstrated activity in cellular and in vivo models.
  • High purity suitable for biochemical and cell-based studies.
  • Soluble in DMSO; available as solid and DMSO solution formats.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-34980 Supplier Medchemexpress LLC Supplier No. HY1262145MG
JH-RE-06 is a small-molecule REV1-REV7 interface inhibitor that disrupts mutagenic translesion synthesis (TLS) by preventing recruitment of POLζ, sensitizing cells to DNA-damaging chemotherapy. It is characterized in cellular and in vivo models for enhancing cisplatin efficacy and reducing therapy-induced mutagenesis; intended for research use only.
  • Targets REV1-REV7 interface to inhibit TLS.
  • Sensitizes tumor cells to DNA-damaging agents.
  • Demonstrated activity in cellular and in vivo models.
  • High purity suitable for biochemical and cell-based studies.
  • Soluble in DMSO; available as solid and DMSO solution formats.
  • Intended for research use only; not for human or clinical use.