Learn More
Medchemexpress LLC JH-RE-06-d6 | 00-00-0 | 98.0% | 474.75 g/mol | C20H10D6Cl3N3O4 | 5 MG

Supplier: Medchemexpress LLC HY126214S5MG
JH-RE-06-d6 is the deuterium-labeled analog of JH-RE-06 supplied for research use to probe REV1-REV7 interface inhibition and translesion synthesis (TLS). It functions as a small-molecule inhibitor that disrupts REV1-REV7 interactions (IC50 = 0.78 μM; Ki = 0.42 μM) and is used in cellular and biochemical studies of POLζ-mediated mutagenic TLS.
- Deuterium-labeled analog useful for tracer or metabolic studies.
- High purity: 98.04%.
- Molecular weight: 474.75 g/mol.
- Chemical formula: C20H10D6Cl3N3O4.
- Appearance: off-white to light yellow solid.
- Storage: powder stable at -20°C; in solvent -80°C (6 months), -20°C (1 month).
- Available in small research sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.