Learn More
Medchemexpress LLC Jh-re-06-d6 | 1361227-90-8 | 98.0% | 474.75 | 1 MG
SDP
Catalog No. 5000436754
Encompass_Preferred
JH-RE-06-d6 is a deuterium labeled compound, derived from JH-RE-06. It functions as a potent inhibitor of the REV1-REV7 interface, targeting REV1 and disrupting mutagenic translesion synthesis. This action helps prevent the recruitment of mutagenic POLζ, thereby improving chemotherapy efficacy.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Targets REV1 to disrupt mutagenic translesion synthesis
  • Improves chemotherapy outcomes
Catalog No. 50-004-36754 Supplier Medchemexpress LLC Supplier No. HY126214S1MG
JH-RE-06-d6 is a deuterium labeled compound, derived from JH-RE-06. It functions as a potent inhibitor of the REV1-REV7 interface, targeting REV1 and disrupting mutagenic translesion synthesis. This action helps prevent the recruitment of mutagenic POLζ, thereby improving chemotherapy efficacy.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Targets REV1 to disrupt mutagenic translesion synthesis
  • Improves chemotherapy outcomes