Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Jh-viii-157-02 | 1639422-97-1 | 98.98% | 465.55 | 25 MG
SDP

Catalog No. 5000439629
Encompass_Preferred

JH-VIII-157-02 is a structural analogue of alectinib, functioning as an ALK inhibitor. It exhibits an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells. It also potently inhibits various EML4-ALK mutants and shows selectivity at other kinases.

  • Inhibits EML4-ALK G1202R (IC50, 2 nM).
  • Potent inhibition of EML4-ALKwt, EAC1156Y, EAF1174L, EAS1206Y (IC50, 2 nM), EAG1269A (IC50, 3 nM), EAL1196M (IC50, 58 nM), EA1151Tins (IC50, 107 nM), and EAL1152R (IC50, 196 nM).
  • Selective inhibition of other kinases including IRAK1, CLK4, RET, RET V804L, RET V804M and IRAK 4.
  • Inhibits growth in cancer cell lines like H3122 and DFCI76 (L1152R) with EC50s of 5 and 19 nM.
  • Good oral bioavailability at 10 mg/kg in mice.
  • Penetrates the CNS of mice.

Catalog No. 50-004-39629 Supplier Medchemexpress LLC Supplier No. 25MGHY112140
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH-VIII-157-02 is a structural analogue of alectinib, functioning as an ALK inhibitor. It exhibits an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells. It also potently inhibits various EML4-ALK mutants and shows selectivity at other kinases.

  • Inhibits EML4-ALK G1202R (IC50, 2 nM).
  • Potent inhibition of EML4-ALKwt, EAC1156Y, EAF1174L, EAS1206Y (IC50, 2 nM), EAG1269A (IC50, 3 nM), EAL1196M (IC50, 58 nM), EA1151Tins (IC50, 107 nM), and EAL1152R (IC50, 196 nM).
  • Selective inhibition of other kinases including IRAK1, CLK4, RET, RET V804L, RET V804M and IRAK 4.
  • Inhibits growth in cancer cell lines like H3122 and DFCI76 (L1152R) with EC50s of 5 and 19 nM.
  • Good oral bioavailability at 10 mg/kg in mice.
  • Penetrates the CNS of mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.