Learn More
Medchemexpress LLC JH-VIII-157-02 | 1639422-97-1 | 99.0% | 465.55 | 50 MG
SDP
Catalog No. 5000437155
Encompass_Preferred
JH-VIII-157-02 is a structural analogue of alectinib and functions as an Anaplastic Lymphoma Kinase (ALK) inhibitor. It demonstrates potent inhibitory activity against various EML4-ALK mutations and shows selectivity at other kinases. The compound also exhibits inhibitory growth against several cancer cell lines and good oral bioavailability in vivo.
  • Potent ALK inhibitor
  • Inhibits various EML4-ALK mutations
  • Selective inhibition against kinases such as IRAK1, CLK4, and RET
  • Inhibits growth of cancer cell lines like H3122 and DFCI76 (L1152R)
  • Good oral bioavailability and CNS penetration
  • Soluble in DMSO at concentrations ≥ 25 mg/mL
Catalog No. 50-004-37155 Supplier Medchemexpress LLC Supplier No. HY11214050MG
JH-VIII-157-02 is a structural analogue of alectinib and functions as an Anaplastic Lymphoma Kinase (ALK) inhibitor. It demonstrates potent inhibitory activity against various EML4-ALK mutations and shows selectivity at other kinases. The compound also exhibits inhibitory growth against several cancer cell lines and good oral bioavailability in vivo.
  • Potent ALK inhibitor
  • Inhibits various EML4-ALK mutations
  • Selective inhibition against kinases such as IRAK1, CLK4, and RET
  • Inhibits growth of cancer cell lines like H3122 and DFCI76 (L1152R)
  • Good oral bioavailability and CNS penetration
  • Soluble in DMSO at concentrations ≥ 25 mg/mL