Learn More
Medchemexpress LLC HY-112140 5mg Medchemexpress, JH-VIII-157-02 CAS:1639422-97-1 Purity:>98%
SDP
Catalog No. 501969037
Encompass_Preferred
Medchemexpress, HY-112140 5mg JH-VIII-157-02 CAS:1639422-97-1 JH-VIII-157-02 is a structural analogue of alectinib, acts as an ALK inhibitor, and shows an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9037 Supplier Medchemexpress LLC Supplier No. HY1121405MG
Medchemexpress, HY-112140 5mg JH-VIII-157-02 CAS:1639422-97-1 JH-VIII-157-02 is a structural analogue of alectinib, acts as an ALK inhibitor, and shows an IC50 of 2 nM for echinoderm microtubule-associated protein-like 4-ALK (EML4-ALK) G1202R in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.