Learn More
Medchemexpress LLC JH-X-119-01 | 2227368-54-7 | 99.0% | 452.46 g/mol | C25H20N6O3 | 100 MG
SDP
Catalog No. 5000247527
Encompass_Preferred
JH-X-119-01 is a research-grade, small-molecule inhibitor selective for interleukin-1 receptor-associated kinase 1 (IRAK1) with reported efficacy in an LPS-induced sepsis mouse model. It is provided as a high-purity powder for preclinical cellular and in vivo studies.
  • Potent and selective IRAK1 inhibition for signaling studies.
  • Demonstrated activity in LPS-induced sepsis in mice.
  • High reported purity suitable for biochemical assays and animal work.
  • Available in multiple powder package sizes to suit experiment scale.
  • Stable with recommended cold storage for long-term use.
Catalog No. 50-002-47527 Supplier Medchemexpress LLC Supplier No. HY103017A100MG
JH-X-119-01 is a research-grade, small-molecule inhibitor selective for interleukin-1 receptor-associated kinase 1 (IRAK1) with reported efficacy in an LPS-induced sepsis mouse model. It is provided as a high-purity powder for preclinical cellular and in vivo studies.
  • Potent and selective IRAK1 inhibition for signaling studies.
  • Demonstrated activity in LPS-induced sepsis in mice.
  • High reported purity suitable for biochemical assays and animal work.
  • Available in multiple powder package sizes to suit experiment scale.
  • Stable with recommended cold storage for long-term use.