Learn More
Medchemexpress LLC JH-X-119-01 | 2227368-54-7 | 99.0% | 452.46 g/mol | C25H20N6O3 | 50 MG
SDP
Catalog No. 5000246258
Encompass_Preferred
JH-X-119-01 is a potent, selective small-molecule inhibitor of interleukin-1 receptor-associated kinase 1 (IRAK1) with an in vitro IC50 of 9 nM. It is supplied for research use as a powder or as a 10 mM solution in DMSO, with a reported purity of 99.03% and molecular weight of 452.46 g/mol.
  • Potent IRAK1 inhibition (IC50 9 nM).
  • High reported purity (99.03%).
  • Molecular weight 452.46 g/mol.
  • Available as powder and as 10 mM DMSO solution.
  • Multiple packaging sizes from milligram to gram scale.
Catalog No. 50-002-46258 Supplier Medchemexpress LLC Supplier No. HY103017A50MG
JH-X-119-01 is a potent, selective small-molecule inhibitor of interleukin-1 receptor-associated kinase 1 (IRAK1) with an in vitro IC50 of 9 nM. It is supplied for research use as a powder or as a 10 mM solution in DMSO, with a reported purity of 99.03% and molecular weight of 452.46 g/mol.
  • Potent IRAK1 inhibition (IC50 9 nM).
  • High reported purity (99.03%).
  • Molecular weight 452.46 g/mol.
  • Available as powder and as 10 mM DMSO solution.
  • Multiple packaging sizes from milligram to gram scale.