Learn More
TARGETMOL CHEMICALS INC JH-X-119-01 5MG
SDP
Catalog No. 502875961
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JH-X-119-01 is a highly potent and selective covalent inhibitor of IRAK-1 (Interleukin-1 Receptor-Associated Kinase 1). Purity 97.1%
Catalog No. 50-287-5961 Supplier TARGETMOL CHEMICALS INC Supplier No. T92035MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. JH-X-119-01 is a highly potent and selective covalent inhibitor of IRAK-1 (Interleukin-1 Receptor-Associated Kinase 1). Purity 97.1%