Learn More
Medchemexpress LLC HY-103017 1mg Medchemexpress, JH-X-119-01 (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030171MG
Encompass_Preferred
Medchemexpress, HY-103017 1mg JH-X-119-01 (hydrochloride) CAS:0 JH-X-119-01 hydrochloride is a potent and selective interleukin-1 receptor-associated kinases 1 (IRAK1) inhibitor. JH-X-119-01 hydrochloride ameliorates LPS-induced sepsis in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9519