Learn More
Medchemexpress LLC HY-111518 1mg Medchemexpress, JH-XI-10-02 CAS:2209085-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1115181MG
Encompass_Preferred
Medchemexpress, HY-111518 1mg JH-XI-10-02 CAS:2209085-22-1 JH-XI-10-02 is a potent and selective degrader of CDK8, with an IC50 of 159 nM, based on PROTAC. JH-XI-10-02 causes proteasomal degradation, does not affect CDK8 mRNA levels. JH-XI-10-02 shows no effect on CDK19. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8899