Learn More
Medchemexpress LLC JH-XVI-178 | 2648453-53-4 | 98.9% | 435.91 | 100 MG
SDP
Catalog No. 5000350725
Encompass_Preferred
JH-XVI-178 is a highly efficient and selective inhibitor of CDK8/19, with IC50 values of 1 nM for CDK8 and 2 nM for CDK19. It exhibits a low clearance rate and moderate oral pharmacokinetic properties.
  • Highly efficient and selective CDK8/19 inhibitor
  • IC50 values of 1 nM for CDK8 and 2 nM for CDK19
  • Low clearance rate
  • Moderate oral pharmacokinetic properties
  • Inhibits the phosphorylation of S727-STAT1 in Jurkat cells
  • Tested in mice model via oral administration at 10 mg/kg
Catalog No. 50-003-50725 Supplier Medchemexpress LLC Supplier No. HY139875100MG
JH-XVI-178 is a highly efficient and selective inhibitor of CDK8/19, with IC50 values of 1 nM for CDK8 and 2 nM for CDK19. It exhibits a low clearance rate and moderate oral pharmacokinetic properties.
  • Highly efficient and selective CDK8/19 inhibitor
  • IC50 values of 1 nM for CDK8 and 2 nM for CDK19
  • Low clearance rate
  • Moderate oral pharmacokinetic properties
  • Inhibits the phosphorylation of S727-STAT1 in Jurkat cells
  • Tested in mice model via oral administration at 10 mg/kg