Learn More
Medchemexpress LLC JH-XVI-178 | 2648453-53-4 | MFCD09701325 | 98.9% | 435.91 g/mol | C22H22ClN7O | 5 MG
SDP
Catalog No. 5000220968
Encompass_Preferred
A potent, selective small-molecule inhibitor of cyclin-dependent kinases CDK8 and CDK19 for research use. The compound displays low clearance and moderate oral pharmacokinetic properties and is supplied as a high-purity research powder with recommended storage and solubility guidance.
  • Potent, selective CDK8 and CDK19 inhibition with low-nanomolar IC50s (1 nM and 2 nM).
  • High purity (≈98.9%) suitable for research applications.
  • Supplied as a small research-scale powder (5 mg).
  • Soluble in DMSO at 62.5 mg/mL; sonication recommended.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-20968 Supplier Medchemexpress LLC Supplier No. HY1398755MG
A potent, selective small-molecule inhibitor of cyclin-dependent kinases CDK8 and CDK19 for research use. The compound displays low clearance and moderate oral pharmacokinetic properties and is supplied as a high-purity research powder with recommended storage and solubility guidance.
  • Potent, selective CDK8 and CDK19 inhibition with low-nanomolar IC50s (1 nM and 2 nM).
  • High purity (≈98.9%) suitable for research applications.
  • Supplied as a small research-scale powder (5 mg).
  • Soluble in DMSO at 62.5 mg/mL; sonication recommended.
  • Stable when stored as powder at -20°C for long-term storage.