Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH295 100mg | 1311143-71-1 | 320.35 g/mol | C18H16N4O2 | 100 MG
SDP

Catalog No. 5000229153
Encompass_Preferred

JH295 is a potent, irreversible, and selective NIMA-related kinase 2 (Nek2) inhibitor for research use. It alkylates Nek2 (Cys22) and shows an IC50 of 770 nM. The compound is a yellow to orange solid (C18H16N4O2, MW 320.35 g/mol) with reported purity ≈98.7%; store at -20°C under nitrogen and prepare fresh solutions due to instability.

  • Selective Nek2 inhibition via covalent alkylation
  • IC50 ~770 nM in biochemical assays
  • Soluble in DMSO (~55 MG/ML); ultrasonic assistance may be required
  • High purity suitable for research (~98.7%)
  • Store at -20°C under inert gas; unstable in solution

Catalog No. 50-002-29153 Supplier Medchemexpress LLC Supplier No. HY116423100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH295 is a potent, irreversible, and selective NIMA-related kinase 2 (Nek2) inhibitor for research use. It alkylates Nek2 (Cys22) and shows an IC50 of 770 nM. The compound is a yellow to orange solid (C18H16N4O2, MW 320.35 g/mol) with reported purity ≈98.7%; store at -20°C under nitrogen and prepare fresh solutions due to instability.

  • Selective Nek2 inhibition via covalent alkylation
  • IC50 ~770 nM in biochemical assays
  • Soluble in DMSO (~55 MG/ML); ultrasonic assistance may be required
  • High purity suitable for research (~98.7%)
  • Store at -20°C under inert gas; unstable in solution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.