Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH295 50mg | 1311143-71-1 | 50 MG
SDP

Catalog No. 5000227989
Encompass_Preferred

JH295 is an irreversible, cysteine-targeted inhibitor of NIMA-related kinase 2 (Nek2) used for biochemical and cellular studies. It irreversibly alkylates Cys22, shows an IC50 of approximately 770 nM, and contains an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).

  • Irreversible Nek2 inhibitor; IC50 ≈ 770 nM.
  • Cysteine-targeted covalent mechanism (alkylation of Cys22).
  • Contains an alkyne handle for click chemistry applications.
  • Molecular weight 320.35; formula C18H16N4O2.
  • High reported purity (98.66%).
  • Solid, yellow to orange appearance.

Catalog No. 50-002-27989 Supplier Medchemexpress LLC Supplier No. HY11642350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH295 is an irreversible, cysteine-targeted inhibitor of NIMA-related kinase 2 (Nek2) used for biochemical and cellular studies. It irreversibly alkylates Cys22, shows an IC50 of approximately 770 nM, and contains an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).

  • Irreversible Nek2 inhibitor; IC50 ≈ 770 nM.
  • Cysteine-targeted covalent mechanism (alkylation of Cys22).
  • Contains an alkyne handle for click chemistry applications.
  • Molecular weight 320.35; formula C18H16N4O2.
  • High reported purity (98.66%).
  • Solid, yellow to orange appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.