Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC JH295 5mg | 1311143-71-1 | 320.35 g/mol | C18H16N4O2 | 5 MG
SDP

Catalog No. 5000227148
Encompass_Preferred

JH295 is a potent, irreversible, and selective NIMA-related kinase 2 (Nek2) inhibitor that contains an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry). Supplied as a 5 mg quantity at 98.66% purity, the compound has a molecular weight of 320.35 g/mol and formula C18H16N4O2; reported cellular IC50 for Nek2 is 770 nM and the mechanism involves covalent alkylation of Cys22.

  • Irreversible Nek2 inhibitor with reported IC50 of 770 nM.
  • Contains an alkyne group for CuAAc click chemistry.
  • Mechanism involves covalent alkylation of cysteine residue Cys22.
  • Supplied as a 5 mg quantity at 98.66% purity.
  • Molecular weight 320.35 g/mol; chemical formula C18H16N4O2.

Catalog No. 50-002-27148 Supplier Medchemexpress LLC Supplier No. HY1164235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JH295 is a potent, irreversible, and selective NIMA-related kinase 2 (Nek2) inhibitor that contains an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry). Supplied as a 5 mg quantity at 98.66% purity, the compound has a molecular weight of 320.35 g/mol and formula C18H16N4O2; reported cellular IC50 for Nek2 is 770 nM and the mechanism involves covalent alkylation of Cys22.

  • Irreversible Nek2 inhibitor with reported IC50 of 770 nM.
  • Contains an alkyne group for CuAAc click chemistry.
  • Mechanism involves covalent alkylation of cysteine residue Cys22.
  • Supplied as a 5 mg quantity at 98.66% purity.
  • Molecular weight 320.35 g/mol; chemical formula C18H16N4O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.